Please login to your online account to display your discounted pricing

JNJ 5207852 dihydrochloride, Tocris™

High affinity H3 receptor antagonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material JNJ 5207852 dihydrochloride
Assay Percent Range >99%
Molecular Formula C20H32N2O·2HCl
Formula Weight 389.4
Synonym 1-[3-[4-(1-Piperidinylmethyl)phenoxy]propyl]piperidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

402010

 
r & d systems
RDS402010
10mg Each for $225.77

402050

 
r & d systems
RDS402050
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 389.4
Quantity 10mg
Chemical Name or Material JNJ 5207852 dihydrochloride
Assay Percent Range >99%
Molecular Formula C20H32N2O·2HCl
CAS 398473-34-2
Synonym 1-[3-[4-(1-Piperidinylmethyl)phenoxy]propyl]piperidine hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature