Please login to your online account to display your discounted pricing

JNJ 5207787, Tocris™

Selective NPY Y2 receptor antagonist

$328.50 - $1,336.15

Chemical Identifiers

Chemical Name or Material JNJ 5207787
Assay Percent Range >98%
Molecular Formula C32H38N4O2
Formula Weight 510.67
Synonym (2E)-N-(1-Acetyl-2, 3-dihydro-1H-indol-6-yl)-3-(3-cyanophenyl)-N-[1-(2-cyclopentylethyl)-4-piperidinyl]-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

401850

 
r & d systems
RDS401850
50mg Each for $1,336.15

401810

 
r & d systems
RDS401810
10mg Each for $328.50
Description & Specifications

Specifications

Formula Weight 510.67
Quantity 50mg
Chemical Name or Material JNJ 5207787
Assay Percent Range >98%
Molecular Formula C32H38N4O2
CAS 683746-68-1
Synonym (2E)-N-(1-Acetyl-2, 3-dihydro-1H-indol-6-yl)-3-(3-cyanophenyl)-N-[1-(2-cyclopentylethyl)-4-piperidinyl]-2-propenamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature