Please login to your online account to display your discounted pricing

JNJ 28871063 hydrochloride, Tocris

Potent ErbB receptor family inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material JNJ 28871063 hydrochloride
Assay Percent Range >97%
Molecular Formula C24H27ClN6O3·HCl
Formula Weight 519.42
Synonym 5E-4-Amino-6-(4-benzyloxy-3-chlorophenylamino)pyrimidine-5-carboxaldehyde N-(2-morpholin-4-ylethyl) oxime hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

335250

 
r & d systems
335250
50mg Each for $1,050.42

335210

 
r & d systems
335210
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 519.42
Quantity 50mg
Chemical Name or Material JNJ 28871063 hydrochloride
Assay Percent Range >97%
Molecular Formula C24H27ClN6O3·HCl
CAS 944342-90-9
Synonym 5E-4-Amino-6-(4-benzyloxy-3-chlorophenylamino)pyrimidine-5-carboxaldehyde N-(2-morpholin-4-ylethyl) oxime hydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C