missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

JNJ 17203212, Tocris

Reversible, competitive and potent TRPV1 antagonist

$212.57 - $889.32

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C17H15F6N5O
Formula Weight 419.32
Synonym 4-[3-(Trifluoromethyl)-2-pyridinyl]-N-[5-(trifluoromethyl)-2-pyridinyl]-1-piperazinecarboxamide
Chemical Name or Material JNJ 17203212
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
336110 R & D Systems
336110
10mg Each for $212.57
336150 R & D Systems
336150
50mg Each for $889.32
Specifications Description & Specifications

Chemical Identifiers

>99%
419.32
JNJ 17203212
C17H15F6N5O
4-[3-(Trifluoromethyl)-2-pyridinyl]-N-[5-(trifluoromethyl)-2-pyridinyl]-1-piperazinecarboxamide

Specifications

10mg
>99%
419.32
JNJ 17203212
821768-06-3
C17H15F6N5O
4-[3-(Trifluoromethyl)-2-pyridinyl]-N-[5-(trifluoromethyl)-2-pyridinyl]-1-piperazinecarboxamide

Safety and Handling

Recommended Storage : Store at +4°C