missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

JNJ 10191584 maleate, Tocris

Selective H4 receptor antagonist; orally active

$176.10 - $741.11

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C13H15ClN4O·C4H4O4
Formula Weight 394.81
Synonym 1-[(5-Chloro-1H-benzimidazol-2-yl)carbonyl]-4-methylpiperazine maleate
Chemical Name or Material JNJ 10191584 maleate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
244150 R & D Systems
244150
50mg Each for $741.11
244110 R & D Systems
244110
10mg Each for $176.10
Specifications Description & Specifications

Chemical Identifiers

>99%
394.81
JNJ 10191584 maleate
C13H15ClN4O·C4H4O4
1-[(5-Chloro-1H-benzimidazol-2-yl)carbonyl]-4-methylpiperazine maleate

Specifications

50mg
>99%
394.81
JNJ 10191584 maleate
869497-75-6
C13H15ClN4O·C4H4O4
1-[(5-Chloro-1H-benzimidazol-2-yl)carbonyl]-4-methylpiperazine maleate

Safety and Handling

Recommended Storage : Desiccate at Room Temperature