Please login to your online account to display your discounted pricing

Alfa Aesar™ JM Special, Multi-element Oil Based Standard, Specpure™, 900μg/g

Chemical Identifiers

Chemical Name or Material JM Special Multi-element Oil Based Standard
Name Note 900μg/g
Purity Grade Specpure™
MDL Number MFCD00151263
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3676622

  SDS
alfa aesar™
3676622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100g
Physical Form Liquid
Chemical Name or Material JM Special Multi-element Oil Based Standard
Name Note 900μg/g
Purity Grade Specpure™
MDL Number MFCD00151263
Concentration Matrix: Hydrocarbon oil
TSCA TSCA