Please login to your online account to display your discounted pricing

Alfa Aesar™ JM-21 Multi-element Oil Based Standard, Specpure™, 900μg/g

Chemical Identifiers

Chemical Name or Material JM-21 Multi-element Oil Based Standard
Name Note 900μg/g
Purity Grade Specpure™
MDL Number MFCD00151262
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3676922

 
alfa aesar™
3676922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material JM-21 Multi-element Oil Based Standard
Quantity 100g
TSCA TSCA
Name Note 900μg/g
Purity Grade Specpure™
Physical Form Liquid
Concentration Matrix: Hydrocarbon oil
MDL Number MFCD00151262