Please login to your online account to display your discounted pricing

JLK 6, Tocris

Inhibitor of γ-secretase-mediated βAPP processing

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material JLK 6
Assay Percent Range >99%
Molecular Formula C10H8ClNO3
Formula Weight 225.63
Synonym 7-Amino-4-chloro-3-methoxy-1H-2-benzopyran
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

267710

 
r & d systems
RDS267710
10mg Each for $192.67

267750

 
r & d systems
RDS267750
50mg Each for $812.12
Description & Specifications

Specifications

Formula Weight 225.63
Quantity 10mg
Molecular Formula C10H8ClNO3
Chemical Name or Material JLK 6
Assay Percent Range >99%
CAS 62252-26-0
Synonym 7-Amino-4-chloro-3-methoxy-1H-2-benzopyran
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature