Please login to your online account to display your discounted pricing

JK 184, Tocris

Alcohol dehydrogenase 7 inhibitor; inhibits downstream Hh signaling pathway

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material JK 184
Assay Percent Range >99%
Molecular Formula C19H18N4OS
Formula Weight 350.44
Synonym N-(4-Ethoxyphenyl)-4-(2-methylimidazo[1, 2-a]pyridin-3-yl)-2-thiazolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

334110

 
r & d systems
RDS334110
10mg Each for $214.12

334150

 
r & d systems
RDS334150
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 350.44
Quantity 10mg
Molecular Formula C19H18N4OS
Assay Percent Range >99%
CAS 315703-52-7
Chemical Name or Material JK 184
Synonym N-(4-Ethoxyphenyl)-4-(2-methylimidazo[1, 2-a]pyridin-3-yl)-2-thiazolamine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C