Please login to your online account to display your discounted pricing

Tocris™ JIB 04

Pan Jumonji inhibitor; active in vivo

$205.70 - $862.95

Specifications

Product Type JIB 04
Inhibitors JIB 04
Molecular Weight 308.76
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

497210R

 
tocris™
RDS497210
10mg Each for $205.70

497250R

 
tocris™
RDS497250
50mg Each for $862.95
Description & Specifications

Specifications

Product Type JIB 04
Inhibitors JIB 04
Molecular Weight 308.76
Purity >99

Pan Jumonji histone demethylase inhibitor (IC50 values are 230, 340, 435, 445, 855 and 1100nM for JARID1A, JMJD2E, JMJD2B, JMJD2A, JMJD3 and JMJD2C respectively). Exhibits negligible activity at PHD2 and TET1. Selectively blocks cancer cell growth in vitro and diminishes tumor growth in H358 and A549 mouse xenograft models in vivo. Prolongs survival in a mouse model of breast cancer.

  • Chemical Name: 5-Chloro-2-[(E)-2-[phenyl(pyridin-2-yl)methylidene]hydrazin-1-yl]pyridine
  • Formula: C17H13ClN4
  • CAS Number: 199596-05-9
  • Solubility: Soluble to 100mM in DMSO and to 50mM in 1eq. HCl
  • Purity: >99