Please login to your online account to display your discounted pricing

Thermo Scientific™ Jewett™ High-Performance Lab Freezers


Store reagents, pharmaceuticals, biologicals and other commonly used laboratory materials.

Manufacturer: thermo scientific™  JLF5030D

 View more versions of this product

Catalog No. JLF5030D

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Warranty Warranty varies by country, please contact us for details
Type High-Performance Freezer
Shelves 8
Shipping Weight (Metric) 272kg
Temperature Control Microprocessor
Doors Double, Solid
Capacity (English) 51.1 cu. ft.
Capacity (Metric) 1447L
Height (Metric) Exterior 201.2cm
Depth (English) Interior 52.5 in.
Width (English) Interior 29 in.
Height (Metric) Interior 147.3cm
Electrical Requirements 208 to 230V 60Hz
Temperature Range (Metric) -30°C
Voltage 208 to 230V
Shipping Weight (English) 600 lb.
Refrigerant CFC-free
Plug Type 6-20P
Height (English) Exterior 79.2 in.
Depth (Metric) Interior 133.4cm
Cabinet Type Upright cabinet
Breaker 15A
Defrost Automatic
Length (English) Exterior 56.5 in.
Capacity 51.1 cu. ft., 1447L
Hertz 60Hz
Width (Metric) Interior 73.7cm
Width (English) Exterior 37.2 in.
Length (Metric) Exterior 143.5cm
Width (Metric) Exterior 94.5cm
Certifications/Compliance UL, CE
Amperage 12A
Height (English) Interior 58 in.
Dimensions (L x W x H) Exterior 56.5 x 37.2 x 79.2 in. (143.5 x 94.5 x 201.2cm)
Dimensions (D x W x H) Interior 52.5 x 29 x 58 in. (133.4 x 73.7 x 147.3cm)

Store reagents, pharmaceuticals, biologicals and other commonly used laboratory materials in Thermo Scientific™ Jewett™ High-Performance Lab Freezers. Designed for medical and scientific applications, these high-performance freezers feature microprocessor control, positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration compressors selected for industrial, clinical and scientific use.

Microprocessor Control

  • Microprocessor control system with audio and visual alarms
  • Bright digital temperature displays
  • Alarm silence, ringback and automatic reset functions
  • Setpoint security with key-operated, triple-position master switch that controls power to all systems
  • Audible/visual alarms for high/low temperature and power failure
  • Battery backup and remote alarm connections
  • Temperature is factory-preset at -30°C.†

Construction

  • Rugged, long-life cabinets with smooth, scratch-resistant interiors‡ and exteriors that are easy to clean
  • High-density, CFC-free insulation
  • Solid self-closing doors with 90° stay-open feature, keyed lock for added sample security
  • Closed-cell gasket provides a positive door seal to maintain a constant interior temperature
  • Directed airflow for fast temperature recovery after door openings
  • Smooth, ergonomic full-length door handle
  • Painted open-wire shelves are adjustable in 1″ (2.5cm) increments
  • 1″ (2.5cm) insulated access port standard
  • Set of four 2″ (5.2cm) casters included, two locking and two regular

Refrigeration System

  • Heavy-duty, industrial-grade, hermetically sealed compressors
  • Automatic defrost maintains optimum cooling

Ordering Information:

Options include freestanding low-voltage booster and built-in seven-day temperature chart recorder. Contact your Customer Service Representative for more information. V and W models not available in North America.

Note:

† Performance may be affected by ambient temperature and humidity conditions, sample loading preferences, and frequency of door openings.

‡ Specified interior volumes are nominal, and usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions..