missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human JAK3, GST Tag Recombinant Protein DFS Item

JAK3 Recombinant Human Protein

$784.00 - $11220.00

Specifications

Accession Number NP_000206
Gene Alias JAK3, LJAK, JAKL
Species Human
Protein Tag GST Tag
Expression System Baculovirus
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV3855 DFS Item
View Documents Promotion Details
Thermo Fisher Scientific
PV3855
10μg
Each for $784.00
 
PV4080 DFS Item
View Documents Promotion Details
Thermo Fisher Scientific
PV4080
1mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Janus kinase 3 (JAK3) phosphorylates members of the STAT (signal transduction and activators of transcription) family. JAK3 is predominantly expressed in immune cells and transduces a signal in response to activation of interleukin receptors.
Specifications

Specifications

NP_000206
Human
Baculovirus
JAK3, LJAK, JAKL
GST Tag
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.