missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Jackson Safety™ G60 Level 5 Cut Resistant Gloves with Dyneema Fiber

$249.74 - $249.74

Specifications

Color Black,Salt and Pepper
Cut Resistance Rating ANSI Level 4/EN388 Level 5
Material Fiber,Polyurethane,Spandex™,Stainless Steel
For Use With (Application) Metal fabrication, Glass handling, handling sharp parts, automotive assembly
Series G60
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Glove Cuff Color Size Price Quantity  
Catalog Number Mfr. No. Glove Cuff Color Size Price Quantity  
17-986-022 Kimberly-Clark™ Professional
KC98237
Purple Large Dozen of 12 Pair for $249.74
Specifications Description & Specifications

Specifications

Black,Salt and Pepper
Fiber,Polyurethane,Spandex™,Stainless Steel
G60
ANSI Level 4/EN388 Level 5
Metal fabrication, Glass handling, handling sharp parts, automotive assembly