Please login to your online account to display your discounted pricing

(±)-J 113397, Tocris

Potent and selective NOP antagonist

$335.85 - $1,384.48

Chemical Identifiers

Chemical Name or Material (±)-J 113397
Assay Percent Range >98%
Molecular Formula C24H37N3O2
Formula Weight 399.57
Synonym (±)-1-[(3R™, 4R™)-1-(Cyclooctylmethyl)-3-(hydroxymethyl)-4-piperidinyl]-3-ethyl-1, 3-dihydro-2H-benzimidazol-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

259810

 
r & d systems
RDS259810
10mg Each for $335.85

259850

 
r & d systems
RDS259850
50mg Each for $1,384.48
Description & Specifications

Specifications

Formula Weight 399.57
Quantity 10mg
CAS 217461-40-0
Molecular Formula C24H37N3O2
Assay Percent Range >98%
Chemical Name or Material (±)-J 113397
Synonym (±)-1-[(3R™, 4R™)-1-(Cyclooctylmethyl)-3-(hydroxymethyl)-4-piperidinyl]-3-ethyl-1, 3-dihydro-2H-benzimidazol-2-one
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature