Please login to your online account to display your discounted pricing

IWP 2, Tocris™

Inhibitor of Wnt processing and secretion; blocks β-catenin accumulation

$214.12 - $214.12

Chemical Identifiers

Chemical Name or Material IWP 2
Assay Percent Range >98%
Molecular Formula C22H18N4O2S3
Formula Weight 466.6
Synonym N-(6-Methyl-2-benzothiazolyl)-2-[(3,4,6,7-tetrahydro-4-oxo-3-phenylthieno[3,2-d]pyrimidin-2-yl)thio]-acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

35-331-0

 
r & d systems
353310
10mg Each for $214.12
Description & Specifications

Specifications

Formula Weight 466.6
Quantity 10mg
Molecular Formula C22H18N4O2S3
Chemical Name or Material IWP 2
Assay Percent Range >98%
CAS 686770-61-6
Synonym N-(6-Methyl-2-benzothiazolyl)-2-[(3,4,6,7-tetrahydro-4-oxo-3-phenylthieno[3,2-d]pyrimidin-2-yl)thio]-acetamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C