Please login to your online account to display your discounted pricing

Ivachtin, Tocris

Potent caspase-3 inhibitor

$133.98 - $279.45

Chemical Identifiers

Chemical Name or Material Ivachtin
Assay Percent Range >99%
Molecular Formula C20H21N3O7S
Formula Weight 447.46
Synonym 2-[4-Methyl-8-(morpholin-4-ylsulfonyl)-1,3-dioxo-1,3-dihydro-2H-pyrrolo[3,4-c]quinolin-2-yl]ethyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

263610

 
r & d systems
RDS263610
10mg Each for $279.45

26361

 
r & d systems
RDS26361
1mg Each for $133.98
Description & Specifications

Specifications

Formula Weight 447.46
Quantity 10mg
Chemical Name or Material Ivachtin
Assay Percent Range >99%
Molecular Formula C20H21N3O7S
CAS 745046-84-8
Synonym 2-[4-Methyl-8-(morpholin-4-ylsulfonyl)-1,3-dioxo-1,3-dihydro-2H-pyrrolo[3,4-c]quinolin-2-yl]ethyl acetate
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C