Texwipe™ Non-Sterile Isopropyl Alcohol
Supplier:  Texwipe™ TX117
 View more versions of this product
Catalog No. 19-130-711
Description
Description
Specifications
Specifications
Isopropyl Alcohol
70% by volume USP-grade Isopropanol and 30% USP purified water
Ideal for surface cleaning, use on gloved hands, wipe down for pass through to controlled environments, use with cleanroom wipers, replaces solvent blending operations in facilities
3.8 L
Liquid
0.2 μm
128.4 oz.