ITK Recombinant Human Protein DFS Item
ITK Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4193
 View more versions of this product
Catalog No. PV4193
Description
Description
ITK is a tyrosine kinase expressed in T-cells which contains both SH2 and SH3 domains. ITK is thought to play a role in T-cell proliferation and differentiation.
Specifications
Specifications
NP_005537
Protein assays and analysis
Frozen
ITK Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
ITK, EMT, LYK, PSCTK2
ITK was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 500μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
ITK Recombinant Human Protein
Human
0.39mg/mL
Frozen
99.9kDa
90% by SDS-PAGE
–80°C
1 Each
ITK Recombinant Human Protein
Kinase assay
TEC
200nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2.08μg/mL
GST Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.