missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ITK Mouse, Clone: 2F12, eBioscience™

Mouse Monoclonal Antibody

$146.52 - $376.20

Specifications

Antigen ITK
Clone 2F12
Host Species Murine
Gene Alias IL-2 inducible T-cell kinase
Applications Immunohistochemistry (Formalin/Paraffin),Microscopy,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122821 Life Technologies
LS14975280
25μg Each for $146.52
501122822 Life Technologies
LS14975282
100μg Each for $376.20
Specifications

Description

The monoclonal antibody 2F12 recognizes human, mouse, and rat ITK. ITK (also known as Interleukin-2 inducible T-cell kinase, T-cell-specific kinase, Tyrosine-protein kinase Lyk, Emt, and Tsk) is a member of the Tec family of kinases that include Tec, Btk, Bmx, and Txk. Tec family kinases play an indispensable role in activation of phospholipase C and activation of calcium-dependent signaling cascades following antigen receptor triggering. ITK is highly expressed by T cells and is required for optimal TCR-induced PLCγ1 activation. Mice congenitally lacking ITK show profound defects in T cell development and signaling. Defects in ITK are the cause of lymphoproliferative syndrome EBV-associated autosomal type 1 (LPSA1). ITK also plays a role in the development of various T cell mediated inflammatory disorders such as atopic dermatitis, psoriasis, and allergic asthma.
Description & Specifications

Specifications

ITK
Murine
Immunohistochemistry (Formalin/Paraffin),Microscopy,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
2F12
IL-2 inducible T-cell kinase
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer