Please login to your online account to display your discounted pricing

ITC Surgicutt™ Bleeding Time Device

Standardized incision depth avoids deep cuts and ensures an incision well above concentrated pain fibers

$662.94 - $714.94

Specifications

Blade Material Stainless Steel
Disposable Yes
For Use With (Application) Avoids deep cuts and ensures an incision well above concentrated pain fibers, used to screen for platelet function
Lancet Style Retractable blade
View More Specs
Products
Catalog Number Mfr. No. Depth (Metric) Length (Metric) Size Product Type Price Quantity    

22-211-507

 
surgilance inc
SUB50I
0.1cm 0.5cm Adult Bleeding Time Device with blotting paper Pack of 50 Each for $714.94

22-211-506

 
surgilance inc
SU501
0.1cm 0.5cm Adult Bleeding Time Device Pack of 50 Each for $662.94
Description & Specifications

Specifications

Blade Material Stainless Steel
Disposable Yes
For Use With (Application) Avoids deep cuts and ensures an incision well above concentrated pain fibers, used to screen for platelet function
Lancet Style Retractable blade

  • Unique, Surgicutt technology provides a uniform incision, not a puncture, improving test standardization and reproducibility of bleeding time test results
  • Stainless-steel blade is a permanent retractable blade—safe and avoids disease transmission
  • Controlled depth and length of incision every time: large blade slot surface reduces skin indentation
  • Virtually painless—diminished bruising means less trauma and more comfort for patients
  • Color coded, disposable, and easy to use
  • Available in three sizes: adult, pediatric, and infant