Please login to your online account to display your discounted pricing

Itaconic anhydride, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  412940250

 View more versions of this product

Catalog No. AC412940250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 112.08
Physical Form Crystalline Powder
Quantity 25g
Color White to cream
Beilstein 17, 442
Melting Point 66° to 70°C
CAS 98684
Infrared Spectrum Authentic
Molecular Formula C5H4O3
Assay Percent Range 98%
Chemical Name or Material Itaconic anhydride
Packaging Glass bottle
MDL Number MFCD00005530
Synonym 2-Methylenesuccinic anhydride
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Itaconic anhydride
CAS 98684
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H4O3
Formula Weight 112.08
MDL Number MFCD00005530
Synonym 2-Methylenesuccinic anhydride
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents