missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Itaconic acid, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 122810010
Specifications
Itaconic acid | |
Authentic | |
1.57 | |
0.1% max. | |
Beige to White | |
1kg | |
99+% | |
HO2CCH2C(=CH2)CO2H | |
15,529 | |
Solubility in water: 95g/L (25°C). Other solubilities: soluble in alcohol and acetone,slightly soluble in most other common organic,solvents | |
C=C(CC(=O)O)C(=O)O | |
130.1 | |
CHEBI:30838 | |
Crystalline Powder or Crystals |
1.5700g/mL | |
Plastic bottle | |
300ppm max | |
0.5% max. | |
165.0°C to 168.0°C | |
97-65-4 | |
C5H6O4 | |
02,76 | |
2-methylenebutanedioic acid, 2-methylenesuccinic acid, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, itaconate, itaconic acid, methylenebutanedioic acid, methylenesuccinic acid, propylenedicarboxylic acid, succinic acid, methylene | |
LVHBHZANLOWSRM-UHFFFAOYSA-N | |
2-methylidenebutanedioic acid | |
811 | |
130.1 | |
99+% |
Chemical Identifiers
97-65-4 | |
130.1 | |
2-methylenebutanedioic acid, 2-methylenesuccinic acid, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, itaconate, itaconic acid, methylenebutanedioic acid, methylenesuccinic acid, propylenedicarboxylic acid, succinic acid, methylene | |
CHEBI:30838 | |
C=C(CC(=O)O)C(=O)O |
C5H6O4 | |
LVHBHZANLOWSRM-UHFFFAOYSA-N | |
811 | |
2-methylidenebutanedioic acid |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 202-599-6