missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isoxazolidine Hydrochloride 97.0+%, TCI America™
$283.68 - $847.75
Chemical Identifiers
CAS | 39657-45-9 |
---|---|
Molecular Formula | C3H8ClNO |
Molecular Weight (g/mol) | 109.553 |
MDL Number | MFCD11100465 |
InChI Key | HPIVVOQIZYNVJE-UHFFFAOYSA-N |
PubChem CID | 12304500 |
IUPAC Name | 1,2-oxazolidine;hydrochloride |
SMILES | C1CNOC1.Cl |
Chemical Identifiers
39657-45-9 | |
109.553 | |
HPIVVOQIZYNVJE-UHFFFAOYSA-N | |
1,2-oxazolidine;hydrochloride |
C3H8ClNO | |
MFCD11100465 | |
12304500 | |
C1CNOC1.Cl |
Specifications
39657-45-9 | |
White | |
MFCD11100465 | |
HPIVVOQIZYNVJE-UHFFFAOYSA-N | |
1,2-oxazolidine;hydrochloride | |
12304500 | |
≥97.0% (T) | |
Isoxazolidine Hydrochloride |
127°C | |
C3H8ClNO | |
1 g | |
C1CNOC1.Cl | |
109.553 | |
109.55 | |
Crystalline Powder |
Safety and Handling
TSCA : No