missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isoxazole 99%, ACROS Organics™

Chemical Identifiers

CAS 288-14-2
Assay Percent Range 99%
Molecular Formula C3H3NO
Formula Weight 69.06
MDL Number MFCD00003149
Chemical Name or Material Isoxazole, 99%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC412930100 SDS Thermo Fisher Scientific
412930100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC412930500 SDS Thermo Fisher Scientific
412930500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

288-14-2
C3H3NO
MFCD00003149
99%
69.06
Isoxazole, 99%

Specifications

10g
1.07g/mL
C3H3NO
MFCD00003149
Glass bottle
288-14-2
99%
69.06
27, 14
Isoxazole, 99%

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-018-7

TSCA : TSCA