Please login to your online account to display your discounted pricing

Isoxazole 99%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  412930500

 View more versions of this product

Catalog No. AC412930500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 69.06
Quantity 50g
EINECS Number 206-018-7
TSCA TSCA
Density 1.07g/mL
Beilstein 27, 14
Chemical Name or Material Isoxazole
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Assay Percent Range 99%
Molecular Formula C3H3NO
MDL Number MFCD00003149
CAS 288-14-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material Isoxazole
Name Note 99%
CAS 288-14-2
Assay Percent Range 99%
Molecular Formula C3H3NO
Formula Weight 69.06
MDL Number MFCD00003149
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.

SDS & Safety Documents