Please login to your online account to display your discounted pricing

Alfa Aesar™ Isoxazole, 99%

Chemical Identifiers

Chemical Name or Material Isoxazole
CAS 288-14-2
Molecular Formula C3H3NO
Formula Weight 69.06
MDL Number MFCD00003149
UN Number UN1993
Beilstein 103773
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0160706

  SDS
alfa aesar™
L0160706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 69.06
Chemical Name or Material Isoxazole
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00003149
Beilstein 103773
UN Number UN1993
TSCA TSCA
Molecular Formula C3H3NO
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
CAS 288-14-2
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Assay 99%
Health Hazard 1 Danger
Quantity 5g
Flash Point 8°C (46°F)
Boiling Point 93° to 95°C
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.