missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isoxazole-5-carboxylic acid, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC10301DA
Specifications
Isoxazole-5-carboxylic acid | |
96.5 | |
97% | |
C4H3NO3 | |
1g | |
MIIQJAUWHSUTIT-UHFFFAOYSA-N | |
1,2-oxazole-5-carboxylic acid | |
2060599 | |
97% |
21169-71-1 | |
100.0 | |
Amber glass bottle | |
MFCD00156151 | |
isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h | |
C1=C(ON=C1)C(=O)O | |
113.072 | |
113.07 |