missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isoxazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H66887MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isoxazole-4-boronic acid pinacol ester | |
110°C to 115°C | |
MFCD06657891 | |
4-isoxazoleboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoxazole, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-isoxazole, isoxazol-4-ylboronic acid pinacol ester, isoxazole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, isoxazole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-isooxazolylboronic acid pinacol ester, 4-isoxazoleboronicacidpinacolester | |
B1(OC(C(O1)(C)C)(C)C)C2=CON=C2 | |
195.025 | |
195.02 |
928664-98-6 | |
C9H14BNO3 | |
250 mg | |
LXCICYRNWIGDQA-UHFFFAOYSA-N | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole | |
16414180 | |
97% |
Chemical Identifiers
928664-98-6 | |
195.025 | |
LXCICYRNWIGDQA-UHFFFAOYSA-N | |
16414180 | |
B1(OC(C(O1)(C)C)(C)C)C2=CON=C2 |
C9H14BNO3 | |
MFCD06657891 | |
4-isoxazoleboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoxazole, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-isoxazole, isoxazol-4-ylboronic acid pinacol ester, isoxazole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, isoxazole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-isooxazolylboronic acid pinacol ester, 4-isoxazoleboronicacidpinacolester | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only