missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isovanillin, TCI America™

$93.58 - $800.23

Chemical Identifiers

CAS 621-59-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00003369
InChI Key JVTZFYYHCGSXJV-UHFFFAOYSA-N
Synonym 3-hydroxy-4-methoxy-benzaldehyde, 3-hydroxy-p-anisaldehyde, 3-hydroxyanisaldehyde, 5-formylguaiacol, benzaldehyde, 3-hydroxy-4-methoxy, iso-vanillin, isovanicaline, isovanilin, isovanillin, isovanilline
PubChem CID 12127
IUPAC Name 3-hydroxy-4-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I051925G View Documents Tci America
I051925G
25g Each for $93.58
 
I0519500G View Documents Tci America
I0519500G
500g Each for $800.23
 
Specifications

Chemical Identifiers

621-59-0
152.149
JVTZFYYHCGSXJV-UHFFFAOYSA-N
12127
COC1=C(C=C(C=C1)C=O)O
C8H8O3
MFCD00003369
3-hydroxy-4-methoxy-benzaldehyde, 3-hydroxy-p-anisaldehyde, 3-hydroxyanisaldehyde, 5-formylguaiacol, benzaldehyde, 3-hydroxy-4-methoxy, iso-vanillin, isovanicaline, isovanilin, isovanillin, isovanilline
3-hydroxy-4-methoxybenzaldehyde

Specifications

179°C
114°C
621-59-0
MFCD00003369
JVTZFYYHCGSXJV-UHFFFAOYSA-N
3-hydroxy-4-methoxybenzaldehyde
12127
Crystal-Powder at 20°C
Isovanillin
Undesignated
25g
C8H8O3
3-hydroxy-4-methoxy-benzaldehyde, 3-hydroxy-p-anisaldehyde, 3-hydroxyanisaldehyde, 5-formylguaiacol, benzaldehyde, 3-hydroxy-4-methoxy, iso-vanillin, isovanicaline, isovanilin, isovanillin, isovanilline
COC1=C(C=C(C=C1)C=O)O
152.149
152.15
≥97.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1200

RTECSNumber : CU6540000

TSCA : Yes