missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isovanillic Acid 98.0+%, TCI America™
Supplier: TCI America I02955G
Specifications
Isovanillic Acid | |
253°C | |
MFCD00002507 | |
isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
COC1=C(C=C(C=C1)C(=O)O)O | |
168.148 | |
CHEBI:63798 | |
≥98.0% (HPLC,T) |
645-08-9 | |
C8H8O4 | |
5 g | |
LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
3-hydroxy-4-methoxybenzoic acid | |
12575 | |
168.15 | |
Crystalline Powder |
Chemical Identifiers
645-08-9 | |
168.148 | |
LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
12575 | |
3-hydroxy-4-methoxybenzoic acid |
C8H8O4 | |
MFCD00002507 | |
isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
CHEBI:63798 | |
COC1=C(C=C(C=C1)C(=O)O)O |
Safety and Handling
RTECSNumber : BZ4850000
TSCA : No