missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isovalerophenone, TCI America™

$90.14 - $242.92

Chemical Identifiers

CAS 582-62-7
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.232
MDL Number MFCD00026486
InChI Key HEOVGVNITGAUKL-UHFFFAOYSA-N
Synonym Isobutyl Phenyl Ketone
PubChem CID 68493
IUPAC Name 3-methyl-1-phenylbutan-1-one
SMILES CC(C)CC(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I02965ML View Documents TCI America
I02965ML
5mL Each for $90.14
 
I029625ML View Documents TCI America
I029625ML
25mL Each for $242.92
 
Specifications

Chemical Identifiers

582-62-7
162.232
HEOVGVNITGAUKL-UHFFFAOYSA-N
68493
CC(C)CC(=O)C1=CC=CC=C1
C11H14O
MFCD00026486
Isobutyl Phenyl Ketone
3-methyl-1-phenylbutan-1-one

Specifications

236°C
5mL
C11H14O
Isobutyl Phenyl Ketone
CC(C)CC(=O)C1=CC=CC=C1
162.232
162.23
≥99.0% (GC)
Yellow
582-62-7
MFCD00026486
HEOVGVNITGAUKL-UHFFFAOYSA-N
3-methyl-1-phenylbutan-1-one
68493
Clear Liquid at 20°C
Isovalerophenone
Safety and Handling

Safety and Handling

TSCA : Yes