missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isovaleric anhydride, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 451090250

 View more versions of this product

Catalog No. AC451090250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Isovaleric anhydride
0.9400g/mL
91°C
Glass bottle
MFCD00015051
0.94
Solubility in water: hydrolyses
CC(C)CC(=O)OC(=O)CC(C)C
186.25
186.25
1468-39-9
215°C
94% min
C10H18O3
25mL
isovaleric anhydride, 3-methylbutanoic anhydride, butanoic acid, 3-methyl-, anhydride, butanoic acid, 3-methyl-, 1,1'-anhydride, isovalericanhydride, isovaleriansaureanhydrid, iso-pentanoic anhydride, acmc-209cxm, isovaleric anhydride gc, butanoic acid,3-methyl-,1,1'-anhydride
FREZLSIGWNCSOQ-UHFFFAOYSA-N
3-methylbutanoyl 3-methylbutanoate
73847
95%

Chemical Identifiers

1468-39-9
186.25
FREZLSIGWNCSOQ-UHFFFAOYSA-N
73847
CC(C)CC(=O)OC(=O)CC(C)C
C10H18O3
MFCD00015051
isovaleric anhydride, 3-methylbutanoic anhydride, butanoic acid, 3-methyl-, anhydride, butanoic acid, 3-methyl-, 1,1'-anhydride, isovalericanhydride, isovaleriansaureanhydrid, iso-pentanoic anhydride, acmc-209cxm, isovaleric anhydride gc, butanoic acid,3-methyl-,1,1'-anhydride
3-methylbutanoyl 3-methylbutanoate
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep only in original container.

GHS Signal Word: Danger

EINECSNumber : 215-994-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only