missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isothiazole, TCI America™

$82.50 - $287.23

Chemical Identifiers

CAS 288-16-4
Molecular Formula C3H3NS
Molecular Weight (g/mol) 85.124
MDL Number MFCD00020818
InChI Key ZLTPDFXIESTBQG-UHFFFAOYSA-N
Synonym isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone
PubChem CID 67515
ChEBI CHEBI:35600
IUPAC Name 1,2-thiazole
SMILES C1=CSN=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0982200MG View Documents TCI America
I0982200MG
200mg Each for $82.50
 
I09821G View Documents TCI America
I09821G
1g Each for $287.23
 
Specifications

Chemical Identifiers

288-16-4
85.124
ZLTPDFXIESTBQG-UHFFFAOYSA-N
67515
1,2-thiazole
C3H3NS
MFCD00020818
isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone
CHEBI:35600
C1=CSN=C1

Specifications

114°C
200mg
C3H3NS
1993
ZLTPDFXIESTBQG-UHFFFAOYSA-N
1,2-thiazole
67515
85.12
≥98.0% (GC)
Colorless
288-16-4
MFCD00020818
isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone
C1=CSN=C1
85.124
CHEBI:35600
Clear Liquid at 20°C
Isothiazole
Safety and Handling

Safety and Handling

TSCA : No