missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isothiazole 98.0+%, TCI America™
Supplier: TCI America I0982200MG
Specifications
Isothiazole | |
Colorless | |
C3H3NS | |
200 mg | |
isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone | |
C1=CSN=C1 | |
85.124 | |
CHEBI:35600 | |
≥98.0% (GC) |
288-16-4 | |
114°C | |
MFCD00020818 | |
1993 | |
ZLTPDFXIESTBQG-UHFFFAOYSA-N | |
1,2-thiazole | |
67515 | |
85.12 | |
Liquid |
Chemical Identifiers
288-16-4 | |
85.124 | |
ZLTPDFXIESTBQG-UHFFFAOYSA-N | |
67515 | |
1,2-thiazole |
C3H3NS | |
MFCD00020818 | |
isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone | |
CHEBI:35600 | |
C1=CSN=C1 |
Safety and Handling
TSCA : No