missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Isotemp™ General Purpose Upright Refrigerator/Freezer
Ideal for storing nonvolatile reagents, samples, and specimens
Manufacturer: Fisherbrand™ 18LCEEFSA
Description
- Temperature range from 1° to 10°C (refrigerator), -12° to -20°C (freezer)
- Total capacity:18.0 cu. ft. (515.4L)
- Refrigerator capacity: 12.9 cu. ft. (365L)
- Freezer capacity: 5.0 cu. ft. (142L)
- Automatic defrost
- Controls at front of refrigerator for easy access
- Separate adjustment of refrigerator and freezer temperatures
- CFC-free R134-A refrigerant
- Yellow warning label reads: “Laboratory Refrigerator” Foamed polyurethane insulation ensures temperature uniformity
- Textured white enamel steel exterior resists marking
- Refrigerator has two white vinyl-coated–steel slide-out adjustable shelves, two tinted slide-out bottom drawers, four door shelves
- Freezer has one compartment and two door shelves
- Two adjustable, two fixed rollers on bottom for easy installation and leveling
Warranty
13 months, parts and labor
Specifications
Refrigerator: 2 adjustable shelves, 1 fixed shelf, 2 drawers, 3 door shelves Freezer: 1 adjustable shelf, 2 door shelves |
|
18 cu. ft. | |
60 Hz | |
Refrigerator: 24.75” X 26.0” X 39.75” (62.86 X 66.04 X 100.9 cm) Freezer : 21.0” X 26.0” X 15.5” (53.34 X 66.04 X 39.4 cm) |
|
93 kg |
cCSAus | |
115 V | |
30.13” X 30” X 66.63” (76.5 X 76.2 X 169.24 cm) | |
205 lb. | |
Refrigerator/Freezer |