missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators, Solid-Stainless Steel Door
Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.
$7,841.83 - $8,815.70
Specifications
Access Security | Key Lock |
---|---|
Certifications/Compliance | cULus,ENERGYSTAR |
Door Style | Solid Stainless Steel |
Electrical Requirements | 115 V 60 Hz |
Plug Type | NEMA 5-15 P |
Catalog Number | Mfr. No. | Capacity (English) | Shelves | Color | Interior | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Shelves | Color | Interior | Price | Quantity | ||
FBG25RPSA
|
Fisherbrand™
FBG25RPSA |
23 cu. ft. | 4 Full | White | Painted White Steel |
Each for $7,841.83
|
|
FBG25RSSA
|
Fisherbrand™
FBG25RSSA |
23 cu. ft. | 4 Full | Gray | Stainless Steel |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
FBG30RPSA
|
Fisherbrand™
FBG30RPSA |
27 cu. ft. | 4 Full | White | Painted White Steel |
Each for $8,815.70
|
|
FBG30RSSA
|
Fisherbrand™
FBG30RSSA |
27 cu. ft. | 4 Full | Gray | Stainless Steel |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
FBG49RPSA
|
Fisherbrand™
FBG49RPSA |
49 cu. ft. | 8 Full | White | Painted White Steel |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
FBG49RSSA
|
Fisherbrand™
FBG49RSSA |
49 cu. ft. | 8 Full | Gray | Stainless Steel |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Whisper-quiet operation
- ENERGY STAR certified
- Temperature, door ajar, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
Specifications
Key Lock | |
Solid Stainless Steel | |
NEMA 5-15 P | |
2°C to 10°C | |
45 kg | |
RPSA | |
5°C |
cULus,ENERGYSTAR | |
115 V 60 Hz | |
R290 | |
100 lb. | |
Microprocessor | |
1 Rear access port | |
2 years parts and labor, plus 3 year compressor warranty |