missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators
Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.
Manufacturer: Fisherbrand™ FBG49RPGA
Description
- Whisper-quiet operation
- ENERGY STAR certified
- Temperature, door ajar, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
Specifications
2°C to 10°C | |
49 cu. ft. | |
115 V 60 Hz | |
NEMA 5-15 P | |
28.5 x 49 x 61.75 in. (724 x 1245 x 1569 mm) | |
271 kg | |
Painted White Steel | |
45 kg | |
2 years parts and labor, plus 3 year compressor warranty | |
5°C | |
1 Rear access port | |
Glass | |
8 Full |
cULus,ENERGYSTAR | |
1387 L | |
8.8 A | |
32 x 52.7 x 81 in.(813 x 1339 x 2057 mm) | |
597 lb. | |
R290 | |
100 lb. | |
Key Lock | |
White | |
RPGA | |
Microprocessor | |
2 |