missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators
Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.
Manufacturer: Fisherbrand™ FBG25RSGA
Description
- Whisper-quiet operation
- ENERGY STAR certified
- Temperature, door ajar, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
Specifications
2°C to 10°C | |
23 cu. ft. | |
115 V 60 Hz | |
NEMA 5-15 P | |
28.5 x 24 x 61.75 in. (724 x 610 x 1569 mm) | |
175 kg | |
Stainless Steel | |
45 kg | |
2 years parts and labor, plus 3 year compressor warranty | |
5°C | |
1 Rear access port | |
Glass | |
4 Full |
cULus,ENERGYSTAR | |
651 L | |
5.8 A | |
32 x 27.75 x 81 in.(813 x 705 x 2057 mm) | |
392 lb. | |
R290 | |
100 lb. | |
Key Lock | |
Gray | |
RSGA | |
Microprocessor | |
1 |