missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.

Manufacturer:  Fisherbrand™ FBG12RPGA

 View more versions of this product

Catalog No. FBG12RPGA



Description

Description

  • Whisper-quiet operation
  • ENERGY STAR certified
  • Temperature, door ajar, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
Specifications

Specifications

2°C to 10°C
12 cu. ft.
115 V 60 Hz
NEMA 5-15 P
20.75 x 21 x 45.75 in.(527 x 533 x 1162 mm)
105 kg
Painted White Steel
45 kg
2 years parts and labor, plus 3 year compressor warranty
5°C
1 Rear access port
Glass
4 Full
cULus,ENERGYSTAR
339 L
5 A
25 x 24.75 x 75.8 in.(635 x 629 x 1926 mm)
230 lb.
R290
100 lb.
Key Lock
White
RPGA
Microprocessor
1
SDS
Documents
Product Certifications
Promotions

Promotions