Fisherbrand™ Isotemp Flammable Materials Freezer
For storing volatile materials, pharmaceuticals, clinical reagents, and biological samples
Manufacturer:  Fisherbrand™ 05FFEEFSA
 View more versions of this product
Catalog No. 05FFEEFSA
Description
Description
  • Accidental ignition of refrigerator contents is prevented by specially designed compressor with enclosed overload, relay, and hydraulic thermostat
  • Static condenser free of electric fan
  • Aluminum evaporator coil
  • Two fixed shelves, three door shelves
  • Requires extra clearance when recessed
Specifications
Specifications
5 cu. ft.
19 x 18.63 x 29.5 in. (48.2 x 47.32 x 74.9cm)
15 in. Upper; 10 in. Bottom
Two fixed shelves, three door shelves
25.75 x 21.38 x 33.13 in. (84.15 x 54.3 x 65.4cm)
-20 to -15°C
05FFEEFSA
Manual
Documents