missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Isotemp™ Explosion Proof Under Counter Freezer

For storing volatile materials in areas with explosive atmospheres or in hazardous locations

Manufacturer:  Fisherbrand™ 05EFEEFSA

 View more versions of this product

Catalog No. 05EFEEFSA



Description

Description

  • Wiring totally enclosed in sealed pipes
  • Hydraulic thermostat enclosed by precision-machined housing
  • Two fixed shelves, three door shelves
  • Must be hardwired to an appropriately rated conduit box
Specifications

Specifications

Undercounter
NFPA,OSHA,UL
141.6 L
29.75 x 21 x 33.5 in. (74.9 x 54.6 x 85.1 cm)
R134a, 100% CFC Free
Explosion-Proof Freezer
6
-15°C to -20°C
5 cu. ft.
115 V 60 Hz
Upper: 15 x 18 x 28 in. (38.1 x 45.7 x 71.1 cm)
Lower: 10 x 18 x 28 in. (25.4 x 45.7 x 71.1 cm)
Manual
CFC-free
SDS
Documents
Product Certifications
Promotions

Promotions