Please login to your online account to display your discounted pricing

Isosorbide, 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC351915000

 View more versions of this product

Catalog No. AC351915000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals, Crystalline Solid or Mass
Formula Weight 146.14
Quantity 500g
Color Off-white to light yellow or beige
Specific Rotation Condition + 42.00 (20.00°C c=3,H2O)
Melting Point 56° to 62°C
Merck Index 15, 5270
Molecular Formula C6H10O4
Chemical Name or Material Isosorbide
CAS 652-67-5
Assay Percent Range 98%
MDL Number MFCD00064827
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Rotation +42° to +48° (20°C, 589nm) (c=3, H2O)

Specifications

Chemical Name or Material Isosorbide
CAS 652-67-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H10O4
Formula Weight 146.14
MDL Number MFCD00064827