missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isosorbide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 351910050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isosorbide | |
652-67-5 | |
5 g | |
Glass bottle | |
97.5% min. (GC) | |
MFCD00064827 | |
isosorbide, isobide, devicoran, hydronol, ismotic, sorbid, +-d-isosorbide, vascardin dinitrate, 3r,3ar,6s,6ar-hexahydrofuro 3,2-b furan-3,6-diol, 1,4-dianhydrosorbitol | |
C1C(C2C(O1)C(CO2)O)O | |
146.14 | |
146.14 |
56.0°C to 62.0°C | |
Beige or White to Yellow | |
Authentic | |
Crystals, Crystalline Solid or Mass | |
C6H10O4 | |
15, 5270 | |
KLDXJTOLSGUMSJ-JGWLITMVSA-N | |
(3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol | |
12597 | |
98% |
Chemical Identifiers
652-67-5 | |
146.14 | |
KLDXJTOLSGUMSJ-JGWLITMVSA-N | |
12597 | |
C1C(C2C(O1)C(CO2)O)O |
C6H10O4 | |
MFCD00064827 | |
isosorbide, isobide, devicoran, hydronol, ismotic, sorbid, +-d-isosorbide, vascardin dinitrate, 3r,3ar,6s,6ar-hexahydrofuro 3,2-b furan-3,6-diol, 1,4-dianhydrosorbitol | |
(3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol |
Safety and Handling
EINECSNumber : 211-492-3
RUO – Research Use Only