Please login to your online account to display your discounted pricing

Alfa Aesar™ Isoquinoline N-oxide, min 50% w/v in water

$69.46 - $209.45

Chemical Identifiers

Chemical Name or Material Isoquinoline N-oxide
CAS 1532-72-5
Molecular Formula C9H7NO
Formula Weight 145.16
MDL Number MFCD00006903
Beilstein 114345
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1479414

  SDS
alfa aesar™
L1479414
25g Each for $209.45

AAL1479406

  SDS
alfa aesar™
L1479406
5g Each for $69.46
Description & Specifications

Specifications

Chemical Name or Material Isoquinoline N-oxide
Concentration ≥50% w/v in water
Odor Odorless
Quantity 25g
CAS 1532-72-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C9H7NO
Formula Weight 145.16
MDL Number MFCD00006903
Beilstein 114345
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.