Please login to your online account to display your discounted pricing

Alfa Aesar™ Isoquinoline N-oxide, min 50% w/v in water


Quantity 25g

Manufacturer: alfa aesar™  L1479414

 View more versions of this product

Catalog No. AAL1479414

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 145.16
Chemical Name or Material Isoquinoline N-oxide
CAS 1532-72-5
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Warning
Molecular Formula C9H7NO
MDL Number MFCD00006903
Beilstein 114345
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 25g
Odor Odorless
Concentration ≥50% w/v in water

Specifications

Chemical Name or Material Isoquinoline N-oxide
CAS 1532-72-5
Molecular Formula C9H7NO
Formula Weight 145.16
MDL Number MFCD00006903
Beilstein 114345
Accessories

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents