missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isoquinoline N-oxide, 98%, Thermo Scientific™
$125.30 - $125.30
Chemical Identifiers
CAS | 1532-72-5 |
---|---|
Molecular Formula | C9H7NO |
Molecular Weight (g/mol) | 145.161 |
MDL Number | MFCD00006903 |
InChI Key | RZIAABRFQASVSW-UHFFFAOYSA-N |
Synonym | isoquinoline n-oxide, isoquinoline 2-oxide, isoquinoline-n-oxide, isoquinoline oxide, isoquinoline, 2-oxide, isoquinolin-n-oxide, isoquinolin-2-ium-2-olate, isoquinolinen-oxide, acmc-1boxv, 2-oxido-isoquinolin-2-ium |
PubChem CID | 290378 |
IUPAC Name | 2-oxidoisoquinolin-2-ium |
SMILES | C1=CC=C2C=[N+](C=CC2=C1)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1973806
|
Thermo Scientific Chemicals
A1973806 |
5 g |
Each for $125.30
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1532-72-5 | |
145.161 | |
RZIAABRFQASVSW-UHFFFAOYSA-N | |
290378 | |
C1=CC=C2C=[N+](C=CC2=C1)[O-] |
C9H7NO | |
MFCD00006903 | |
isoquinoline n-oxide, isoquinoline 2-oxide, isoquinoline-n-oxide, isoquinoline oxide, isoquinoline, 2-oxide, isoquinolin-n-oxide, isoquinolin-2-ium-2-olate, isoquinolinen-oxide, acmc-1boxv, 2-oxido-isoquinolin-2-ium | |
2-oxidoisoquinolin-2-ium |
Specifications
1532-72-5 | |
170°C (3 mmHg) | |
MFCD00006903 | |
114345 | |
isoquinoline n-oxide, isoquinoline 2-oxide, isoquinoline-n-oxide, isoquinoline oxide, isoquinoline, 2-oxide, isoquinolin-n-oxide, isoquinolin-2-ium-2-olate, isoquinolinen-oxide, acmc-1boxv, 2-oxido-isoquinolin-2-ium | |
C1=CC=C2C=[N+](C=CC2=C1)[O-] | |
145.161 | |
145.16 | |
Isoquinoline N-oxide |
103°C to 105°C | |
C9H7NO | |
5 g | |
Hygroscopic | |
RZIAABRFQASVSW-UHFFFAOYSA-N | |
2-oxidoisoquinolin-2-ium | |
290378 | |
98% |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 216-242-7
TSCA : No
Recommended Storage : Ambient temperatures