Please login to your online account to display your discounted pricing

Alfa Aesar™ Isoquinoline-5-carboxylic acid, 96%

$167.26 - $464.08

Chemical Identifiers

Chemical Name or Material Isoquinoline-5-carboxylic acid
CAS 27810-64-6
Molecular Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD03788744
Beilstein 129180
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1963703

  SDS
alfa aesar™
L1963703
1g Each for $464.08

AAL19637MD

  SDS
alfa aesar™
L19637MD
250mg Each for $167.26
Description & Specifications

Specifications

Formula Weight 173.17
MDL Number MFCD03788744
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H7NO2
CAS 27810-64-6
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Chemical Name or Material Isoquinoline-5-carboxylic acid
Health Hazard 1 Warning
Melting Point 278° to 281°C
Beilstein 129180
Quantity 1g
Boiling Point 60° to 62°C (11mmHg)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.