missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isoquinoline-4-carboxylic acid, 97%, Maybridge

Chemical Identifiers

CAS 7159-36-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD00094344
Chemical Name or Material Isoquinoline-4-carboxylic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC04901CB SDS Fisher Scientific Company
CC04901CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC04901DA SDS Fisher Scientific Company
CC04901DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7159-36-6
100.0
C10H7NO2
MFCD00094344
97%
96.5
97%
173.17
Isoquinoline-4-carboxylic acid

Specifications

250mg
96.5
97%
173.17
Amber Glass Bottle
97%
7159-36-6
100.0
C10H7NO2
MFCD00094344
Isoquinoline-4-carboxylic acid

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning