missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isoquinoline-3-carboxylic Acid Monohydrate, TCI America™

$389.59 - $1,223.78

Chemical Identifiers

CAS 203626-75-9
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00075137
InChI Key IXFLTVWYYQIVRB-UHFFFAOYSA-N
Synonym isoquinoline-3-carboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate, isoquinoline-3-carboxylic acid monohydrate, isoquinoline-3-carboxylic acid hydrate 1:x, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, pubchem10167, acmc-1clhb, 3-isoquinolinecarboxylicacid hydrate, isoquinoline 3-carboxylic acid hydrate
PubChem CID 2736704
IUPAC Name isoquinoline-3-carboxylic acid;hydrate
SMILES C1=CC=C2C=NC(=CC2=C1)C(=O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I05095G View Documents TCI America
I05095G
5g Each for $1,223.78
 
I05091G View Documents TCI America
I05091G
1g Each for $389.59
 
Specifications

Chemical Identifiers

203626-75-9
191.186
IXFLTVWYYQIVRB-UHFFFAOYSA-N
2736704
C1=CC=C2C=NC(=CC2=C1)C(=O)O.O
C10H9NO3
MFCD00075137
isoquinoline-3-carboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate, isoquinoline-3-carboxylic acid monohydrate, isoquinoline-3-carboxylic acid hydrate 1:x, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, pubchem10167, acmc-1clhb, 3-isoquinolinecarboxylicacid hydrate, isoquinoline 3-carboxylic acid hydrate
isoquinoline-3-carboxylic acid;hydrate

Specifications

Undesignated
5g
C10H9NO3
isoquinoline-3-carboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate, isoquinoline-3-carboxylic acid monohydrate, isoquinoline-3-carboxylic acid hydrate 1:x, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, pubchem10167, acmc-1clhb, 3-isoquinolinecarboxylicacid hydrate, isoquinoline 3-carboxylic acid hydrate
C1=CC=C2C=NC(=CC2=C1)C(=O)O.O
191.186
173.17
≥98.0% (HPLC,T)
167°C
203626-75-9
MFCD00075137
IXFLTVWYYQIVRB-UHFFFAOYSA-N
isoquinoline-3-carboxylic acid;hydrate
2736704
Crystal-Powder at 20°C
Isoquinoline-3-carboxylic Acid Monohydrate
Safety and Handling

Safety and Handling

TSCA : No