missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isoquinoline-3-carboxylic acid hydrate, 97%, Maybridge™

$112.70 - $112.70

Chemical Identifiers

CAS 203626-75-9
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00075137
InChI Key IXFLTVWYYQIVRB-UHFFFAOYSA-N
Synonym 3-isoquinolinecarboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylicacid hydrate, acmc-1clhb, isoquinoline 3-carboxylic acid hydrate, isoquinoline-3-carboxylic acid hydrate, isoquinoline-3-carboxylic acid hydrate 1:x, isoquinoline-3-carboxylic acid monohydrate, pubchem10167
PubChem CID 2736704
IUPAC Name isoquinoline-3-carboxylic acid;hydrate
SMILES C1=CC=C2C=NC(=CC2=C1)C(=O)O.O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC04701DA
SDS View Documents
Fisher Scientific Company
CC04701DA
1g Amber Glass Bottle
Each for $112.70
 
Description

Description

Specifications

Chemical Identifiers

203626-75-9
191.186
IXFLTVWYYQIVRB-UHFFFAOYSA-N
2736704
C1=CC=C2C=NC(=CC2=C1)C(=O)O.O
C10H9NO3
MFCD00075137
3-isoquinolinecarboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylicacid hydrate, acmc-1clhb, isoquinoline 3-carboxylic acid hydrate, isoquinoline-3-carboxylic acid hydrate, isoquinoline-3-carboxylic acid hydrate 1:x, isoquinoline-3-carboxylic acid monohydrate, pubchem10167
isoquinoline-3-carboxylic acid;hydrate

Specifications

Amber Glass Bottle
203626-75-9
100.0
C10H9NO3
3-isoquinolinecarboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylicacid hydrate, acmc-1clhb, isoquinoline 3-carboxylic acid hydrate, isoquinoline-3-carboxylic acid hydrate, isoquinoline-3-carboxylic acid hydrate 1:x, isoquinoline-3-carboxylic acid monohydrate, pubchem10167
C1=CC=C2C=NC(=CC2=C1)C(=O)O.O
191.186
191.19
Isoquinoline-3-carboxylic acid hydrate
1g
96.5
97%
MFCD00075137
IXFLTVWYYQIVRB-UHFFFAOYSA-N
isoquinoline-3-carboxylic acid;hydrate
2736704
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS